Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRR20A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571242
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571242 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571242 Supplier Invitrogen™ Supplier No. PA571242
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Human: 100%.

This gene is one of five identical loci in a cluster on chromosome 13q21.1. The predicted protein is proline-rich and contains several dopamine D4 receptor signatures and PRINTS domains.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRR20A
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene PRR20A
Gene Accession No. P86496
Gene Alias proline rich 20; proline rich 20A; proline-rich protein 20; proline-rich protein 20A; Proline-rich protein 20B; Proline-rich protein 20C; Proline-rich protein 20D; Proline-rich protein 20E; PRR20; PRR20A; PRR20B; PRR20C; PRR20D; PRR20E
Gene Symbols PRR20A
Host Species Rabbit
Immunogen synthetic peptide directed towards the N-terminal region of human PRR20A.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 122183
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.