Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PRR23A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA583867
Description
Immunogen sequence: CASVPEIAAQ EEAYEEDADP EFPEL.
PRR23A is a protein coding gene.
Specifications
PRR23A | |
Polyclonal | |
Unconjugated | |
PRR23A | |
proline rich 23A; proline-rich protein 23A; proline-rich protein 23C; PRR23A; UPF0572 protein ENSP00000372650 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
729627 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
A6NEV1 | |
PRR23A | |
Recombinant protein corresponding to Human PRR23A. Recombinant protein control fragment (Product #RP-108175). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction