Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRR26 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584705
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584705 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584705 Supplier Invitrogen™ Supplier No. PA584705
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SPTPPKLSPG QLSPHSVNVH WGPQGHLHLP RSGTTVLHAY LQTLSSPAS Highest antigen sequence indentity to the following orthologs: Rat - 33%, Mouse - 34%.

PRR26 is a protein coding gene. Diseases associated with PRR26 include Hepatocellular Carcinoma.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRR26
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PRR26
Gene Accession No. Q8N8Z3
Gene Alias C10orf108; FLJ38681; proline rich 26; PRR26
Gene Symbols PRR26
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PRR26. Recombinant protein control fragment (Product #RP-103619).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 414235
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.