Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRR3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566660
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566660 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566660 Supplier Invitrogen™ Supplier No. PA566660
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LSLPPPPWGRG PIRRGLGPRS SPYGRGWWGV NAEPPFPGPG HGGPTRGSFH KEQRNPRRLK SWSLIKNTCP PKDDPQVMED KSDRPVCRH Highest antigen sequence identity to the following orthologs - mouse 86%, rat 83%.

PRR3 gene ontology annotations related to this gene include metal ion binding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRR3
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PRR3
Gene Accession No. P79522
Gene Alias CAT56; MHC class I region proline-rich protein CAT56; proline rich 3; proline-rich polypeptide 3; proline-rich protein 3; PRR3
Gene Symbols PRR3
Host Species Rabbit
Immunogen Recombinant Human PRR3. Recombinant protein control fragment (Product #RP-107308).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80742
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.