Learn More
Invitrogen™ PRR4 Monoclonal Antibody (1D2)
GREENER_CHOICE
Mouse Monoclonal Antibody
Supplier:  Invitrogen™ MA524440
Catalog No. PIMA524440
Only null left
Add to Cart
Description
Description
Peptide Sequence: QSTDNDVNYE DFTFTIPDVE DSSQRPDQGP QRPPPEGLLP RPPGDSGNQD DGPQQRPPKP GGHHRHPPPP PFQNQQRPPQ RGHRQLSLPR FPSVSLQEAS.
Lacrimal proline rich protein is a member of the proline-rich protein family which lacks a conserved repetitive domain. It may have a role in protective functions in the eye. Two alternatively spliced transcript variants that encode different proteins have been described for this gene.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PRR4
Monoclonal
Unconjugated
PRR4
Lacrimal proline-rich protein; LPRP; Nasopharyngeal carcinoma-associated proline-rich protein 4; PROL4; proline rich 4 (lacrimal); proline-rich polypeptide 4; proline-rich protein 4; PRR4
Mouse
Protein A
RUO
11272
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot
1D2
PBS with no preservative; pH 7.4
Q16378
PRR4
PRR4 (NP_009175.1, 17 a.a. ~ 116 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
100 μg
Primary
Human
Antibody
IgG2a κ
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.