Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PRR4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA559883
Description
Immunogen sequence: QSTDNDVNYE DFTFTIPDVE DSSQRPDQGP QRPPPEGLLP RPPGDSGNQD DGPQQRPP Highest antigen sequence identity to the following orthologs: Mouse - 38%, Rat - 38%.
Lacrimal proline rich protein is a member of the proline-rich protein family which lacks a conserved repetitive domain. It may have a role in protective functions in the eye. Two alternatively spliced transcript variants that encode different proteins have been described for this gene.
Specifications
PRR4 | |
Polyclonal | |
Unconjugated | |
PRR4 | |
Lacrimal proline-rich protein; LPRP; Nasopharyngeal carcinoma-associated proline-rich protein 4; PROL4; proline rich 4 (lacrimal); proline-rich polypeptide 4; proline-rich protein 4; PRR4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11272 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q16378 | |
PRR4 | |
Recombinant protein corresponding to Human PRR4. Recombinant protein control fragment (Product #RP-97674). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction