Learn More
Invitrogen™ PRR5 Polyclonal Antibody

Description
Positive test controls include: 293T, HepG2, SH-SY5Y, Mouse liver, Mouse brain, Rat liver. Immunogen sequence: SRGVTEDYLR LETLVQKVVS PYLGTYGLHS SEGPFTHSCI LEKRLLRRSR SGDVLAKNPV VRSKSYNTPL LNPVQEHEAE GAAAGGTSIR RHSVSEMTSC PEPQGFSDPP GQGPTGTFRS SPAPHSGPCP SRLYPTTQPP EQGLDPTRSS LPRSSPENLV DQILESVDSD SEGIFIDFGR GRGSGMSDLE GSGGRQSVV.
Specifications
Specifications
| Antigen | PRR5 |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.89 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 |
| Gene | PRR5 |
| Gene Accession No. | P85299, Q5FVG6, Q812A5 |
| Gene Alias | Arhgap8; AU043908; C030017C09Rik; C78947; FLJ20185k; PP610; proline rich 5; proline rich 5 (renal); proline-rich protein 5; Protein observed with Rictor-1; Protor1; protor-1; prr5; prr5 {ECO:0000250; Rho GTPase activating protein 8; RP1-181C9.3; UniProtKB:P85299} |
| Gene Symbols | PRR5 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.