Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRR5L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567089
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567089 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567089 Supplier Invitrogen™ Supplier No. PA567089
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RQCSSEPNITD NPDGLEEGAR GSQEGSELNC ASLS Highest antigen sequence identity to the following orthologs - mouse 73%, rat 73%.

Proline-rich protein 5-like (PRR5L), also known as Protor-2, is structurally similar to PRR5 and also binds to the mTOR complex 2 (mTORC2). PRR5L is not required for mTORC2 integrity or kinase activity, but dissociates from mTORC2 upon knockdown of tuberous sclerosis complex 1 (TSC1) and TSC2. Knockdown of PRR5L reduced apoptosis in mTORC2 deficient cells as well as cells where mTOR was active, suggesting that mTORC2-dissociated PRR5L may promote apoptosis, and thus play a role in the balance between cell growth and cell death.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRR5L
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PRR5L
Gene Accession No. Q6MZQ0
Gene Alias 2600010E01Rik; 4833411O04Rik; proline rich 5 like; proline-rich protein 5-like; Protein observed with Rictor-2; PROTOR2; protor-2; Prr5l
Gene Symbols PRR5L
Host Species Rabbit
Immunogen Recombinant Human PRR5L. Recombinant protein control fragment (Product #RP-107733).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79899
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.