Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PRR7 Antibody (TRAP3/10), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP130424B
Description
PRR7 Monoclonal antibody specifically detects PRR7 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
PRR7 | |
Monoclonal | |
Biotin | |
PBS | |
Mouse | |
Protein A purified | |
RUO | |
80758 | |
Store at 4C in the dark. | |
IgG2a |
Western Blot, Immunocytochemistry | |
TRAP3/10 | |
Western Blot, Immunocytochemistry/ Immunofluorescence | |
MGC10772, proline rich 7 (synaptic), proline-rich protein 7, Synaptic proline-rich membrane protein | |
Recombinant C-terminal half of the intracellular domain of human PRR7/TRAP3 (amino acids 126-253) | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction