Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRR7 Antibody (TRAP3/10), DyLight 550, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP130424R

Catalog No. NBP130424R


Only null left
Add to Cart

Description

Description

PRR7 Monoclonal specifically detects PRR7 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

PRR7
Monoclonal
DyLight 550
50 mM sodium borate with 0.05% sodium azide
PRR7
Recombinant C-terminal half of the intracellular domain of human PRR7/TRAP3 (amino acids 126-253)
0.1 mL
80758.0
Human, Mouse, Rat
IgG2a
Western Blot, Immunocytochemistry, Immunofluorescence
TRAP3/10
Western Blot, Immunocytochemistry/Immunofluorescence
MGC10772, proline rich 7 (synaptic), proline-rich protein 7, Synaptic proline-rich membrane protein
Mouse
Protein A purified
Primary
The mouse monoclonal antibody TRAP3/10 recognizes an epitope located in the C-terminal part of the intracellular domain of PRR7/TRAP3 (amino acids 126-253 of human PRR7/ TRAP3), a 28 kDa proline-rich membrane protein presumably associated with NMDA receptor complex.
Store at 4°C in the dark.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.