Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRR7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561266
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561266 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561266 Supplier Invitrogen™ Supplier No. PA561266
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MSKPPCYEEA VLMAEPPPPY SEVLTDTRGL YRKIVTPFLS RRDSAEKQEQ PPPSYKPLFL DRGYTSALHL PSAPRPAPPC PALCLQADRG RRVFPSWTDS ELSSREPLEH GAWRLPVSIP Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.

PRR7/TRAP3 (proline-rich 7, transmembrane adaptor protein 3) is a 28 kDa transmembrane adaptor protein ubiquitously expressed at low level (most in brain). Its amino acid sequence is extremely conserved among mammalian and other species. PRR7/TRAP3 contains potential palmitoylation motif and is found in lipid rafts. It is a part of the complex postsynaptic density fraction in neurons and associates with PSD-95, NMDA receptor and probably other proteins. The intracellular domain of PRR7/TRAP3 contains several tyrosines, a proline-rich sequence, and a C-terminal PDZ-binding motif. So far nothing is known about function of this protein. It may be involved in regulation of some receptor signaling and in formation of neurologic and immunologic synapse.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRR7
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PRR7
Gene Accession No. Q8TB68
Gene Alias MGC10772; proline rich 7 (synaptic); proline rich 7, synaptic; proline-rich protein 7; Prr7; Synaptic proline-rich membrane protein; XM_484263
Gene Symbols PRR7
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PRR7. Recombinant protein control fragment (Product #RP-90213).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80758
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.