Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRRC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558698
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558698 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558698 Supplier Invitrogen™ Supplier No. PA558698
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AFQEVFGLAV VVGEAGQSNI APQPVGYAAG LKGAQERIDS LRRTGVIHEK QTAVSVENFI AELLPDKWFD IGCLVVEDPV H Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.

PRRC1 gene ontology annotations related to this gene include activation of protein kinase A activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRRC1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PRRC1
Gene Accession No. Q96M27
Gene Alias 1190002C06Rik; 2310058D16Rik; 3110038B19Rik; 9430085A19Rik; FLJ32875; Prcc1; proline rich coiled-coil 1; proline-rich and coiled-coil-containing protein 1; proline-rich coiled-coil 1; protein PRRC1; Prrc1
Gene Symbols PRRC1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PRRC1. Recombinant protein control fragment (Product #RP-96953).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 133619
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.