Learn More
PRRC1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A305783AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
PRRC1 gene ontology annotations related to this gene include activation of protein kinase A activity.Specifications
PRRC1 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q96M27 | |
PRRC1 | |
between 175 and 225 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunoprecipitation, Western Blot | |
Unconjugated | |
PRRC1 | |
FLJ32875, proline-rich and coiled-coil-containing protein 1, proline-rich coiled-coil 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
133619 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.