Learn More
PRRC1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31
Specifications
| Antigen | PRRC1 | 
|---|---|
| Applications | Immunoprecipitation, Western Blot | 
| Classification | Polyclonal | 
| Conjugate | Unconjugated | 
| Form | Liquid | 
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
| 50-157-1211   | Bethyl Laboratories, Inc A305784AT | 20 μL | 
                                        
                                            
                                                
                                                
                                                
                                            
                                        
                                        
                                            
                                            
                                                
                                                
                                            
                                        
                                        
                                            
                                                
                                                Each of 1 for $151.31
                                                 |  | |||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rat, Zebrafish and Chicken
PRRC1 gene ontology annotations related to this gene include activation of protein kinase A activity.Specifications
| PRRC1 | |
| Polyclonal | |
| Liquid | |
| RUO | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| FLJ32875, proline-rich and coiled-coil-containing protein 1, proline-rich coiled-coil 1 | |
| between 225 and 275 | |
| Primary | |
| 4° C | |
| PRRC1 | 
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| Q3UPH1, Q96M27 | |
| 133619, 73137 | |
| IgG | |
| Antigen affinity chromatography | |
| Antibody | 
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
