Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRRC1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. 501735783 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-5783 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-5783 Supplier Proteintech Group Inc Supplier No. 278871AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PRRC1 gene ontology annotations related to this gene include activation of protein kinase A activity.

Specifications

Antigen PRRC1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene PRRC1
Gene Accession No. Q3UPH1, Q96M27
Gene Alias proline rich coiled coil 1, Protein PRRC1, PRRC1
Gene Symbols Prrc1
Host Species Rabbit
Immunogen PRRC1 Fusion Protein Ag27349
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 133619, 73137
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.