Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PRRC2B Antibody, Biotin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298065B
Description
PRRC2B Polyclonal specifically detects PRRC2B in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
PRRC2B | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
BAT2L, DKFZp781F05101, HLA-B associated transcript 2-like 1, HLA-B-associated transcript 2-like 1, LQFBS-1, MGC10526, proline-rich coiled-coil 2B, proline-rich coiled-coil protein 2B, protein BAT2-like 1, protein PRRC2B | |
Produced in rabbits immunized with E. coli-derived Human PRRC2B fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Biotin | |
PBS | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
84726 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction