Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRRC2B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554309
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554309 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554309 Supplier Invitrogen™ Supplier No. PA554309
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EREESTLKKG DCRDSWRSNK GCSEDHSGLD AKSRGPRAFG RALPPRLSNC GYGRRTFVSK ESPHWQSKSP GSSWQEYGPS DTCGSRRPTD RDYVPDSYRH PDAFGGR Highest antigen sequence identity to the following orthologs: Mouse - 69%, Rat - 71%.

PRRC2B (Proline Rich Coiled-Coil 2B) is a Protein Coding gene. An important paralog of this gene isPRRC2C.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRRC2B
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PRRC2B
Gene Accession No. Q5JSZ5
Gene Alias 5830434P21Rik; AI173903; BAT2L; Bat2l1; D430039P21; HLA-B associated transcript 2-like 1; HLA-B-associated transcript 2-like 1; Kiaa0515; LQFBS-1; mKIAA0515; proline rich coiled-coil 2B; proline-rich coiled-coil 2B; proline-rich coiled-coil protein 2B; protein BAT2-like 1; protein PRRC2B; PRRC2B
Gene Symbols PRRC2B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PRRC2B. Recombinant protein control fragment (Product #RP-92954).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84726
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.