Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRRC2C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555936
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555936 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555936 Supplier Invitrogen™ Supplier No. PA555936
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TTSQSSKQPP PSIRLPSAQT PNGTDYVASG KSIQTPQSHG TLTAELWDNK VAPPAVLNDI SKKLGPISPP QPPSVSAWNK PLTSFGSAP Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 30%.

Proline-rich coiled-coil 2C (PRRC2C) is proposed to be involved in protein C-terminus binding and contains a conserved BAT2 N-terminus domain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRRC2C
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PRRC2C
Gene Accession No. Q9Y520
Gene Alias BAT2 domain containing 1; BAT2 domain-containing protein 1; BAT2D1; BAT2-iso; BAT2L2; HBV XAg-transactivated protein 2; HBV X-transactivated gene 2 protein; HBxAg transactivated protein 2; HLA-B associated transcript 2-like 2; HLA-B-associated transcript 2-like 2; KIAA1096; proline rich coiled-coil 2C; proline-rich and coiled-coil-containing protein 2C; proline-rich coiled-coil 2C; protein BAT2-like 2; protein PRRC2C; PRRC2C; XTP2
Gene Symbols PRRC2C
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PRRC2C. Recombinant protein control fragment (Product #RP-94126).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23215
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.