Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRRC2C Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501564820
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-4820 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-4820 Supplier Bethyl Laboratories, Inc Supplier No. A303314AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

Proline-rich coiled-coil 2C (PRRC2C) is proposed to be involved in protein C-terminus binding and contains a conserved BAT2 N-terminus domain.

Specifications

Antigen PRRC2C
Applications Immunoprecipitation
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene PRRC2C
Gene Accession No. Q9Y520
Gene Alias BAT2 domain containing 1, BAT2 domain-containing protein 1, BAT2D1, BAT2-iso, BAT2L2, HBV XAg-transactivated protein 2, HBV X-transactivated gene 2 protein, HBxAg transactivated protein 2, HLA-B associated transcript 2-like 2, HLA-B-associated transcript 2-like 2, KIAA1096, proline-rich and coiled-coil-containing protein 2C, proline-rich coiled-coil 2C, protein BAT2-like 2, XTP2
Gene Symbols PRRC2C
Host Species Rabbit
Immunogen Between 325 and 375
Purification Method Antigen affinity chromatography
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23215
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.