Learn More
PRRC2C Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A303315A

Description
The recommended shelf life for this product is 1 year from date of receipt.
Proline-rich coiled-coil 2C (PRRC2C) is proposed to be involved in protein C-terminus binding and contains a conserved BAT2 N-terminus domain.Specifications
PRRC2C | |
Polyclonal | |
Unconjugated | |
PRRC2C | |
BAT2 domain containing 1, BAT2 domain-containing protein 1, BAT2D1, BAT2-iso, BAT2L2, HBV XAg-transactivated protein 2, HBV X-transactivated gene 2 protein, HBxAg transactivated protein 2, HLA-B associated transcript 2-like 2, HLA-B-associated transcript 2-like 2, KIAA1096, proline-rich and coiled-coil-containing protein 2C, proline-rich coiled-coil 2C, protein BAT2-like 2, XTP2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23215 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q9Y520 | |
PRRC2C | |
Between 2250 and 2300 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.