Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRRG1 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$487.50

Specifications

Antigen PRRG1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP159907
SDP
View Documents
Novus Biologicals
NBP159907
100 μL
Each of 1 for $487.50
Only null left
Add to Cart
 
Description

Description

PRRG1 Polyclonal specifically detects PRRG1 in Human samples. It is validated for Western Blot.
Specifications

Specifications

PRRG1
Polyclonal
Rabbit
O14668
5638
Synthetic peptides corresponding to PRRG1(proline rich Gla (G-carboxyglutamic acid) 1) The peptide sequence was selected from the N terminal of PRRG1. Peptide sequence ECKEEFCTFEEAREAFENNEKTKEFWSTYTKAQQGESNRGSDWFQFYLTF.
Primary
Western Blot
Unconjugated
RUO
PRGP1TMG1, proline rich Gla (G-carboxyglutamic acid) 1, Proline-rich gamma-carboxyglutamic acid protein 1, proline-rich Gla (G-carboxyglutamic acid) polypeptide 1, Proline-rich Gla protein 1, transmembrane gamma-carboxyglutamic acid protein 1
PRRG1
IgG
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.