Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRRG1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569301
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569301 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569301 Supplier Invitrogen™ Supplier No. PA569301
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 93%; Dog: 93%; Guinea Pig: 86%; Horse: 93%; Human: 100%; Mouse: 93%; Pig: 93%; Rabbit: 93%; Rat: 93%.

This gene encodes a vitamin K-dependent, gamma-carboxyglutamic acid (Gla)-containing, single-pass transmembrane protein. This protein contains a Gla domain at the N-terminus, preceded by a propeptide sequence required for post-translational gamma-carboxylation of specific glutamic acid residues by a vitamin K-dependent gamma-carboxylase. The C-terminus is proline-rich containing PPXY and PXXP motifs found in a variety of signaling and cytoskeletal proteins. This gene is highly expressed in the spinal cord. Several alternatively spliced transcript variants have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRRG1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene PRRG1
Gene Accession No. O14668
Gene Alias 2010007L08Rik; ortholog of proline rich Gla (G-carboxyglutamic acid) 1 PRRG1; PRGP1; proline rich and Gla domain 1; proline rich Gla (G-carboxyglutamic acid) 1; Proline-rich gamma-carboxyglutamic acid protein 1; proline-rich Gla (G-carboxyglutamic acid) polypeptide 1; Proline-rich Gla protein 1; PRRG1; RGD1561320; TMG1; Transmembrane gamma-carboxyglutamic acid protein 1; transmembrane gamma-carboxyglutamic acid protein 1-like
Gene Symbols PRRG1
Host Species Rabbit
Immunogen synthetic peptide directed towards the N terminal region of human PRRG1.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5638
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.