Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PRRG1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569302
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 93%; Pig: 100%; Rabbit: 100%; Rat: 93%.
This gene encodes a vitamin K-dependent, gamma-carboxyglutamic acid (Gla)-containing, single-pass transmembrane protein. This protein contains a Gla domain at the N-terminus, preceded by a propeptide sequence required for post-translational gamma-carboxylation of specific glutamic acid residues by a vitamin K-dependent gamma-carboxylase. The C-terminus is proline-rich containing PPXY and PXXP motifs found in a variety of signaling and cytoskeletal proteins. This gene is highly expressed in the spinal cord. Several alternatively spliced transcript variants have been found for this gene.
Specifications
PRRG1 | |
Polyclonal | |
Unconjugated | |
PRRG1 | |
2010007L08Rik; ortholog of proline rich Gla (G-carboxyglutamic acid) 1 PRRG1; PRGP1; proline rich and Gla domain 1; proline rich Gla (G-carboxyglutamic acid) 1; Proline-rich gamma-carboxyglutamic acid protein 1; proline-rich Gla (G-carboxyglutamic acid) polypeptide 1; Proline-rich Gla protein 1; PRRG1; RGD1561320; TMG1; Transmembrane gamma-carboxyglutamic acid protein 1; transmembrane gamma-carboxyglutamic acid protein 1-like | |
Rabbit | |
Affinity chromatography | |
RUO | |
5638 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
O14668 | |
PRRG1 | |
synthetic peptide directed towards the N terminal region of human PRRG1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction