Learn More
Invitrogen™ PRRG2 Monoclonal Antibody (7D1)

Description
Peptide Sequence: ANHWDLELLT PGNLERECLE ERCSWEEARE YFEDNTLTER FWESYIYNGK GGRGRVDVAS.
Specifications
Specifications
| Antigen | PRRG2 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 7D1 |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | PRRG2 |
| Gene Accession No. | O14669 |
| Gene Alias | 1600014G02Rik; 2510041M06Rik; N4WBP1; NEDD4 WW domain-binding protein 1; PRGP2; proline rich and Gla domain 2; proline rich Gla (G-carboxyglutamic acid) 2; proline-rich gamma-carboxyglutamic acid protein 2; proline-rich Gla (G-carboxglutamic acid) polypeptide 2; proline-rich Gla (G-carboxyglutamic acid) polypeptide 2; proline-rich Gla protein 2; Prrg2; TMG2; Transmembrane gamma-carboxyglutamic acid protein 2 |
| Gene Symbols | PRRG2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.