Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PRRG4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA555110
Description
Immunogen sequence: ARGPKASKHA GEEVFTSKEE ANFFIHRRLL YNRFDLELFT PGNLERECNE ELCNYEEARE IFVDEDKTIA FWQEYSAKGP TTKSDGNR Highest antigen sequence identity to the following orthologs: Mouse - 70%, Rat - 70%.
PRRG4 gene ontology annotations related to this gene include calcium ion binding.
Specifications
PRRG4 | |
Polyclonal | |
Unconjugated | |
PRRG4 | |
9930111I18Rik; hypothetical protein LOC550330; PRGP4; proline rich and Gla domain 4; proline rich Gla (G-carboxyglutamic acid) 4 (transmembrane); proline rich Gla 4 (transmembrane) isoform 1; proline rich Gla 4 (transmembrane) isoform 2; proline rich Gla 4 (transmembrane) isoform 3; proline-rich gamma-carboxyglutamic acid protein 4; Proline-rich Gla protein 4; PRRG4; RGD1560542; TMG4; Transmembrane gamma-carboxyglutamic acid protein 4; zgc:110234 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79056 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BZD6 | |
PRRG4 | |
Recombinant protein corresponding to Human PRRG4. Recombinant protein control fragment (Product #RP-90250). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction