Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRRG4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115801
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115801 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115801 Supplier Invitrogen™ Supplier No. PA5115801
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total PRRG4.

PRRG4 gene ontology annotations related to this gene include calcium ion binding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRRG4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene PRRG4
Gene Accession No. Q8BGN6, Q9BZD6
Gene Alias 9930111I18Rik; hypothetical protein LOC550330; PRGP4; proline rich and Gla domain 4; proline rich Gla (G-carboxyglutamic acid) 4 (transmembrane); proline rich Gla 4 (transmembrane) isoform 1; proline rich Gla 4 (transmembrane) isoform 2; proline rich Gla 4 (transmembrane) isoform 3; proline-rich gamma-carboxyglutamic acid protein 4; Proline-rich Gla protein 4; PRRG4; RGD1560542; TMG4; Transmembrane gamma-carboxyglutamic acid protein 4; zgc:110234
Gene Symbols PRRG4
Host Species Rabbit
Immunogen A synthesized peptide derived from human PRRG4(Accession Q9BZD6), corresponding to amino acid residues E81-F131.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 228413, 499847, 79056
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.