Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PRRS Virus M Protein Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5111971
Description
PRRS Virus M Protein Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
Porcine reproductive and respiratory syndrome virus (PRRSV) is a highly infectious pathogen that causes severe diseases in pigs and great economic losses to the swine industry worldwide. GP5 and M are the major envelope proteins encoded by PRRSV.
Specifications
| PRRS Virus M Protein | |
| Polyclonal | |
| Unconjugated | |
| Porcine reproductive and respiratory syndromevirus | |
| Recombinant protein encompassing a sequence within the center region of PRRS virus M protein (PRRS virus (strain VR-2332)). The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.23 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| Virus | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction