Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PRRS Virus Nsp1 beta Protein Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5112016
Description
PRRS Virus Nsp1 beta Protein Polyclonal Antibody for Western Blot
Porcine reproductive and respiratory syndrome virus (PRRSV) is a highly infectious pathogen that causes severe diseases in pigs and great economic losses to the swine industry worldwide. The viral nonstructural protein 1 (NSP1) of the PRRS virus (PRRSV) is able to suppress the type I interferon (IFN) response in vitro.
Specifications
| PRRS Virus Nsp1 beta Protein | |
| Polyclonal | |
| Unconjugated | |
| Porcine reproductive and respiratory syndromevirus | |
| Full length PRRS virus Nsp1 beta recombinant protein (PRRS virus (strain VR-2332)). | |
| 100 μL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| Virus | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction