Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRRS Virus Nucleocapsid Protein Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117308
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117308 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5117308 Supplier Invitrogen™ Supplier No. PA5117308
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Centrifuge briefly prior to opening.

Betaarterivirus suid 1, formerly (PRRS, Porcine reproductive and respiratory syndrome virus), is a virus that causes a disease of pigs, called porcine reproductive and respiratory syndrome, also known as blue-ear pig disease. The Nucleocapsid protein is an internal structure of the virus that comprised of the association of the N protein and the RNA genome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRRS Virus Nucleocapsid Protein
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Alias Porcine reproductive and respiratory syndrome virus ORF7; PRRSV ORF7
Host Species Rabbit
Immunogen Full length PRRS virus Nucleocapsid recombinant protein (PRRS virus (strain US5)).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.