Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PRRS Virus Nucleocapsid Protein Recombinant Rabbit Monoclonal Antibody (HL2046)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA550366
Description
PRRS Virus Nucleocapsid Protein Recombinant Monoclonal Antibody for Western Blot, ICC/IF
Betaarterivirus suid 1, formerly (PRRS, Porcine reproductive and respiratory syndrome virus), is a virus that causes a disease of pigs, called porcine reproductive and respiratory syndrome, also known as blue-ear pig disease. The Nucleocapsid protein is an internal structure of the virus that comprised of the association of the N protein and the RNA genome.
Specifications
PRRS Virus Nucleocapsid Protein | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
Rabbit | |
Protein A | |
RUO | |
Virus | |
Antibody | |
IgG |
Western Blot, Immunocytochemistry | |
HL2046 | |
Unconjugated | |
Porcine reproductive and respiratory syndrome virus ORF7; PRRSV ORF7 | |
Full length PRRS virus Nucleocapsid protein. | |
100 μL | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction