Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRRS Virus ORF7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117524
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117524 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5117524 Supplier Invitrogen™ Supplier No. PA5117524
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody has been tested in direct-ELISA.

ORF7 is the nucleoprotein gene of porcine reproductive and respiratory syndrome virus (PRRSV). The ORF7 encodes the nucleocapsid protein (N), the most abundant viral protein in virus-infected cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRRS Virus ORF7
Applications Western Blot
Classification Polyclonal
Concentration 2.15 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene Alias ORF7
Host Species Rabbit
Immunogen Recombinant Porcine reproductive and respiratory syndrome virus Nucleocapsid protein (1-123aa).
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.