Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRRSV nsp4 Antibody, Novus Biologicals™
SDP

Catalog No. NB176964 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB176964 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB176964 Supplier Novus Biologicals Supplier No. NBP313277
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PRRSV nsp4 Polyclonal antibody specifically detects PRRSV nsp4 in Virus samples. It is validated for Western Blot

Specifications

Antigen PRRSV nsp4
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS, 20% Glycerol (pH7)
Gene Alias 3C-like serine proteinase, 3CLSP, Non-structural protein 4, Porcine reproductive and respiratory syndrome virus nsp4, Serine protease nsp4
Host Species Rabbit
Immunogen Full length PRRS virus Nsp4 recombinant protein. (PRRS virus (strain VR-2332))
Purification Method Antigen Affinity-purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.