Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRRSV Rabbit anti-Virus, Polyclonal, Bioss
SDP

Catalog No. 501980646
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-0646 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-0646 Supplier Bioss Supplier No. BS10043R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PRRSV belongs to the family Arteriviridae in the genus Arterivirus, order Nidovirales. This group also includes lactate dehydrogenase-elevating virus (LDV) of mice, equine arteritis virus (EAV) and simian hemorrhagic fever virus (SHFV). PRRSV is a small, enveloped virus of 45-70 nm in diameter. Its genome consists of a polyadenylated, single-stranded, non-segmented, positivesense RNA polycystronic molecule of 15.1-15.5 kb. It contains, in the 5 to 3 direction, a 5 leader sequence, at least 10 open reading frames (ORF1a, ORF1b and ORFs 2a, 2b, 3, 4, 5, 5a, 6 and 7) and a 3 non-translated region. ORF1a and ORF1b are located directly downstream of the 5 leader and encode a large replicase polyprotein, which is thought to be autoproteolytically cleaved into at least 13 smaller non-structural proteins including a viral RNA-dependent RNA polymerase.

Specifications

Antigen PRRSV
Applications ELISA, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Host Species Rabbit
Immunogen Derived from an inactive PRRSV vaccine strain NVDC-JXA1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.