Learn More
PRRT1 Mouse anti-Human,Mouse,Rat, Unconjugated, Clone: L102/45, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | PRRT1 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | L102/45 |
| Conjugate | Unconjugated |
| Gene Accession No. | NP_001027456.1 |
| Gene Alias | Proline-rich transmembrane protein 1;Dispanin subfamily D member 1;DSPD1;Synapse differentiation-induced protein 4 |
| Gene Symbols | Prrt1, Ng5 |
| Host Species | Mouse |
| Immunogen | GST-tagged recombinant fragment corresponding to the first 66 amino acids from the N-terminal region of rat Proline-rich transmembrane protein 1 (PRRT1). |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.