Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRRT1 Mouse anti-Human,Mouse,Rat, Unconjugated, Clone: L102/45, MilliporeSigma™

Catalog No. MABN18672MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN18672MI 25 μL
MABN18671MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABN18672MI Supplier MilliporeSigma Supplier No. MABN186725UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PRRT1 Monoclonal antibody specifically detects PRRT1 in Human,Mouse,Rat samples. It is validated for Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRRT1
Applications Western Blot
Classification Monoclonal
Clone L102/45
Conjugate Unconjugated
Gene Accession No. NP_001027456.1
Gene Alias Proline-rich transmembrane protein 1;Dispanin subfamily D member 1;DSPD1;Synapse differentiation-induced protein 4
Gene Symbols Prrt1, Ng5
Host Species Mouse
Immunogen GST-tagged recombinant fragment corresponding to the first 66 amino acids from the N-terminal region of rat Proline-rich transmembrane protein 1 (PRRT1).
Molecular Weight of Antigen ∼40 kDa observed; 31.39 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Protein G Purified
Quantity 25 μL
Research Discipline Neuroscience
Primary or Secondary Primary
Test Specificity Clone L102/45 is a mouse monoclonal antibody that detects Proline-rich transmembrane protein 1 (PRRT1). It targets an epitope within 66 amino acids from the N-terminal, extracellular domain.
Target Species Human, Mouse, Rat
Content And Storage Stable for 1 year at 2 to 8°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.