Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PRRT1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5100309
Description
Antibody detects endogenous levels of total PRRT1.
PRRT1 is a protein coding gene.
Specifications
PRRT1 | |
Polyclonal | |
Unconjugated | |
PRRT1 | |
Bat4; Bat-4; C6orf31; C7H6orf31; D17H6S54E; Dispanin subfamily D member 1; DSPD1; G5b; IFITMD7; interferon induced transmembrane protein domain containing 7; Ng5; Orf31; proline rich transmembrane protein 1; proline-rich transmembrane protein 1; PRRT1; prrt1 {ECO:0000250; Synapse differentiation-induced protein 4; SynDIG4; UniProtKB:Q99946} | |
Rabbit | |
Affinity chromatography | |
RUO | |
260297, 406167, 80863 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O35449, Q6MG82, Q99946 | |
PRRT1 | |
A synthesized peptide derived from human PRRT1(Accession Q99946), corresponding to amino acid residues M1-S39. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction