Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PRRT1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5111480
Description
Immunogen sequence: TLPRLGAGGL ASSAATAQRG PSSSAT.
PRRT1 is a protein coding gene.
Specifications
| PRRT1 | |
| Polyclonal | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q99946 | |
| PRRT1 | |
| Recombinant Protein corresonding to Human PRRT1. Recombinant protein control fragment (Product #RP-108437). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PRRT1 | |
| Bat4; Bat-4; C6orf31; C7H6orf31; D17H6S54E; Dispanin subfamily D member 1; DSPD1; G5b; IFITMD7; interferon induced transmembrane protein domain containing 7; Ng5; Orf31; proline rich transmembrane protein 1; proline-rich transmembrane protein 1; PRRT1; prrt1 {ECO:0000250; Synapse differentiation-induced protein 4; SynDIG4; UniProtKB:Q99946} | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 80863 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction