Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRRT2 Monoclonal Antibody (CL4233)
GREENER_CHOICE

Catalog No. PIMA531398
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531398 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531398 Supplier Invitrogen™ Supplier No. MA531398
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Immunogen sequence: PKPALQPELP TQEDPTPEIL SESVGEKQEN GAVVPLQAGD GEEGPAPEPH SPPSKKSPPA NGAPPRVLQQ LVEEDRMRRA HSGHPGSPRG SLSRHPSSQL AGPGVEGGEG TQKPRDY Highest antigen sequence indentity to the following orthologs: Rat - 87%, Mouse - 88%.

The proline-rich transmembrane protein 2 (PRRT2) contains a proline-rich domain in its N-terminal half and is predominantly expressed in brain and spinal cord in embryonic and postnatal stages. While little is known of the function of this protein, mutations in PRRT2 have been shown to be the causative gene of paroxysmal kinesigenic dyskinesia, which is characterized by recurrent, brief attacks of abnormal involuntary movements induced by sudden voluntary movements. Recent studies have shown that PRRT2 may also be involved in some forms of benign familial infantile epilepsy (BFIE), an autosomal dominant epilepsy syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRRT2
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CL4233
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PRRT2
Gene Accession No. Q7Z6L0
Gene Alias 1500031I19Rik; AI195361; BFIC2; BFIS2; Dispanin subfamily B member 3; DSPB3; dystonia 10; DYT10; EKD1; FICCA; ICCA; IFITMD1; infantile convulsions and paroxysmal choreoathetosis; interferon induced transmembrane protein domain containing 1; PKC; proline rich transmembrane protein 2; proline-rich transmembrane protein 2; PRRT2; RGD1564195
Gene Symbols PRRT2
Host Species Mouse
Immunogen Recombinant protein fragment of Human PRRT2 (Product #RP-91025)
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 112476
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.