Learn More
PRRT2 Mouse anti-Human, Clone: OTI9H8, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF809867

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The proline-rich transmembrane protein 2 (PRRT2) contains a proline-rich domain in its N-terminal half and is predomitly expressed in brain and spinal cord in embryonic and postnatal stages. While little is known of the function of this protein, mutations in PRRT2 have been shown to be the causative gene of paroxysmal kinesigenic dyskinesia, which is characterized by recurrent, brief attacks of abnormal involuntary movements induced by sudden voluntary movements. Recent studies have shown that PRRT2 may also be involved in some forms of benign familial infantile epilepsy (BFIE), an autosomal domit epilepsy syndrome.Specifications
PRRT2 | |
Monoclonal | |
Unconjugated | |
PRRT2 | |
BFIC2, BFIS2, DSPB3, DYT10, EKD1, FICCA, ICCA, IFITMD1, PKC | |
Mouse | |
Affinity Chromatography | |
RUO | |
112476 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI9H8 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q7Z6L0 | |
PRRT2 | |
Human recombit protein fragment corresponding to amino acids 152-268 of human PRRT2 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.