Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PRRX1 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP281848
Description
PRRX1 Polyclonal specifically detects PRRX1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.Specifications
PRRX1 | |
Polyclonal | |
Unconjugated | |
PBS | |
PRRX1 | |
PRRX1 antibody was raised against a 19 amino acid peptide near the center of human PRRX1. The immunogen is located within amino acids 70 - 120 of PRRX1. Amino Acid Squence: DNDQLNSEEKKKRKQRRNR | |
Affinity Purified | |
RUO | |
Primary | |
PRRX1 antibody is human, mouse and rat reactive. At least two isoforms of PRRX1 are known to exist; this antibody will detect both isoforms. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot, ELISA | |
1 mg/mL | |
Western Blot 1-2 ug/mL, ELISA | |
Homeobox protein PHOX1, paired mesoderm homeo box 1, paired mesoderm homeobox 1 isoform pmx-1b, paired mesoderm homeobox protein 1, paired related homeobox 1, Paired-related homeobox protein 1, PHOX1PRX-1, PMX1PRX1 | |
Rabbit | |
26 kDa | |
0.1 mg | |
Growth and Development, Neuronal Cell Markers | |
5396 | |
Human, Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction