Learn More
PRRX1 Mouse anti-Human, Clone: OTI1E10, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF803116

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The DNA-associated protein encoded by this gene is a member of the paired family of homeobox proteins localized to the nucleus. The protein functions as a transcription co-activator, enhancing the DNA-binding activity of serum response factor, a protein required for the induction of genes by growth and differentiation factors. The protein regulates muscle creatine kinase, indicating a role in the establishment of diverse mesodermal muscle types. Alternative splicing yields two isoforms that differ in abundance and expression patterns.Specifications
PRRX1 | |
Monoclonal | |
Unconjugated | |
PRRX1 | |
AGOTC, PHOX1, PMX1, PRX-1, PRX1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
5396 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI1E10 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P54821 | |
PRRX1 | |
Full length human recombit protein of human PRRX1 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.