Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRSS16 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113435
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113435 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113435 Supplier Invitrogen™ Supplier No. PA5113435
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PRSS16 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

This gene encodes a serine protease expressed exclusively in the thymus. It is thought to play a role in the alternative antigen presenting pathway used by cortical thymic epithelial cells during the positive selection of T cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRSS16
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 3.96 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene PRSS16
Gene Accession No. Q9NQE7, Q9QXE5
Gene Alias AI448615; FLJ36271; FLJ40714; FLJ44172; protease, serine 16; protease, serine 16 (thymus); protease, serine, 16; protease, serine, 16 (thymus); Prss16; Serine protease 16; thymus specific serine peptidase; thymus-specific serine protease; Tssp
Gene Symbols PRSS16
Host Species Rabbit
Immunogen Recombinant Human Thymus-specific serine protease protein (352-489AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10279, 364719, 54373
Target Species Human, Mouse, Rat
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.