Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRSS16 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542135
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542135 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542135 Supplier Invitrogen™ Supplier No. PA542135
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: SHSTPYCGLR RAVQIVLHSL GQKCLSFSRA ETVAQLRSTE PQLSGVGDRQ Sequence homology: Cow: 86%; Dog: 100%; Guinea Pig: 85%; Horse: 92%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 93%; Rat: 93%.

This gene encodes a serine protease expressed exclusively in the thymus. It is thought to play a role in the alternative antigen presenting pathway used by cortical thymic epithelial cells during the positive selection of T cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRSS16
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene PRSS16
Gene Accession No. Q9NQE7
Gene Alias AI448615; FLJ36271; FLJ40714; FLJ44172; protease, serine 16; protease, serine 16 (thymus); protease, serine, 16; protease, serine, 16 (thymus); Prss16; Serine protease 16; thymus specific serine peptidase; thymus-specific serine protease; Tssp
Gene Symbols PRSS16
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human PRSS16.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10279
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.