Learn More
Invitrogen™ PRSS27 Monoclonal Antibody (6F11)
Mouse Monoclonal Antibody
Supplier:  Invitrogen™ MA522709
Catalog No. PIMA522709
Only null left
Add to Cart
Description
Description
Peptide Sequence: QLVQPGPHAM YARVRQVESN PLYQGTASSA DVALVELEAP VPFTNYILPV CLPDPSVIFE TGMNCWVTGW GSPSEEDLLP EPRILQKLAV PIIDTPKCNL LYSKDTEFGY.
PRSS27, also known as Marapsin, is a gene located on chromosome 16p13.3 that encodes a serine protease enzyme. This protein is part of the trypsin-like serine protease family, which is characterized by a catalytic triad essential for proteolytic activity. PRSS27 is primarily expressed in the epidermis and other epithelial tissues, such as the salivary glands and pancreas. It plays a role in various physiological processes, including digestion, inflammation, and immune response regulation. In the skin, PRSS27 is involved in keratinocyte differentiation and the maintenance of skin barrier function. Dysregulation or altered expression of PRSS27 has been associated with inflammatory skin conditions, such as psoriasis, where it may influence the inflammatory cascade and contribute to disease pathogenesis. Research into PRSS27 continues to explore its potential role in epithelial homeostasis and as a therapeutic target for skin-related and other inflammatory disorders.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PRSS27
Monoclonal
1 mg/mL
PBS with no preservative; pH 7.4
Q9BQR3
PRSS27
PRSS27 (NP_114154, 94 a.a. ~ 203 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
100 μg
Primary
Human
Antibody
IgG2a κ
ELISA, Western Blot
6F11
Unconjugated
PRSS27
CAP-2; CAPH2; channel-activating protease 2; channel-activating protease 2-like protein; EC 3.4.21; EC 3.4.21.4; EC 3.4.21.43; marapsin; Mpn; MPNMarapsin; Pancreasin; protease, serine 27; PRSS27; Serine protease 27; UNQ1884/PRO4327
Mouse
Affinity Chromatography
RUO
83886
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.