Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRSS27 Monoclonal Antibody (04), Invitrogen™
GREENER_CHOICE

Catalog No. PIMA543032
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA543032 100 μL
PIMA543536 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIMA543032 Supplier Invitrogen™ Supplier No. MA543032
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Specificity: Human Pancreasin/Marapsin/PRSS27

PRSS27, also known as Marapsin, is a gene located on chromosome 16p13.3 that encodes a serine protease enzyme. This protein is part of the trypsin-like serine protease family, which is characterized by a catalytic triad essential for proteolytic activity. PRSS27 is primarily expressed in the epidermis and other epithelial tissues, such as the salivary glands and pancreas. It plays a role in various physiological processes, including digestion, inflammation, and immune response regulation. In the skin, PRSS27 is involved in keratinocyte differentiation and the maintenance of skin barrier function. Dysregulation or altered expression of PRSS27 has been associated with inflammatory skin conditions, such as psoriasis, where it may influence the inflammatory cascade and contribute to disease pathogenesis. Research into PRSS27 continues to explore its potential role in epithelial homeostasis and as a therapeutic target for skin-related and other inflammatory disorders.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRSS27
Applications ELISA
Classification Monoclonal
Clone 04
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene PRSS27
Gene Accession No. Q9BQR3
Gene Alias CAP-2; CAPH2; channel-activating protease 2; channel-activating protease 2-like protein; EC 3.4.21; EC 3.4.21.4; EC 3.4.21.43; marapsin; Mpn; MPNMarapsin; Pancreasin; protease, serine 27; PRSS27; Serine protease 27; UNQ1884/PRO4327
Gene Symbols PRSS27
Host Species Mouse
Immunogen Recombinant Human Pancreasin/Marapsin/PRSS27 Protein.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83886
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.