Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRSS27 Recombinant Rabbit Monoclonal Antibody (107)
GREENER_CHOICE

Catalog No. PIMA529506
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA529506 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA529506 Supplier Thermo Scientific Supplier No. MA529506
Add to Cart
Add to Cart

PRSS27 Recombinant Rabbit Monoclonal Antibody (107)

PRSS27 Recombinant Monoclonal Antibody for Western Blot, IP

TRUSTED_SUSTAINABILITY

Specifications

Antigen PRSS27
Applications Immunoprecipitation, Western Blot
Classification Recombinant Monoclonal
Clone 107
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene PRSS27
Gene Accession No. Q9BQR3
Gene Alias CAP-2; CAPH2; channel-activating protease 2; channel-activating protease 2-like protein; EC 3.4.21; EC 3.4.21.4; EC 3.4.21.43; marapsin; Mpn; MPNMarapsin; Pancreasin; protease, serine 27; PRSS27; Serine protease 27; UNQ1884/PRO4327
Gene Symbols PRSS27
Host Species Rabbit
Immunogen Recombinant Human Pancreasin/Marapsin/PRSS27 protein (Met1-Lys290).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83886
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.