Learn More
Invitrogen™ PRSS3 Monoclonal Antibody (419911)

Description
Reconstitute at 0.5 mg/mL in sterile PBS.
Specifications
Specifications
| Antigen | PRSS3 |
| Applications | Immunohistochemistry (Frozen) |
| Classification | Monoclonal |
| Clone | 419911 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5% trehalose and No Preservative |
| Gene | PRSS3 |
| Gene Accession No. | 0 |
| Gene Alias | Brain trypsinogen; mesotrypsin; Mesotrypsinogen; MTG; pancreatic trypsinogen III; protease, serine 3; protease, serine, 3; protease, serine, 4 (trypsin 4, brain); PRSS3; PRSS4; Serine protease 3; Serine protease 4; T9; Tb; Try3; TRY4; trypsin 3; trypsin III; Trypsin IV; trypsin-3; trypsinogen; trypsinogen 15; trypsinogen 4; trypsinogen 5; trypsinogen IV |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.