Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRSS3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA548034
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA548034 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA548034 Supplier Invitrogen™ Supplier No. PA548034
Add to Cart
Add to Cart

Goat Polyclonal Antibody

In Western blots, approximately 20% cross-reactivity with recombinant human (rh) Trypsin 1, rhTrypsin 2, and rhTrypsin 3 is shown. Reconstitute at 0.2 mg/mL in sterile PBS.

PRSS3 encodes a trypsinogen, which is a member of the trypsin family of serine proteases. This enzyme is expressed in the brain and pancreas and is resistant to common trypsin inhibitors. It is active on peptide linkages involving the carboxyl group of lysine or arginine. PRSS3 is localized to the locus of T cell receptor beta variable orphans on chromosome 9. Four transcript variants encoding different isoforms have been described for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRSS3
Applications Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene PRSS3
Gene Accession No. 0
Gene Alias Brain trypsinogen; mesotrypsin; Mesotrypsinogen; MTG; pancreatic trypsinogen III; protease, serine 3; protease, serine, 3; protease, serine, 4 (trypsin 4, brain); PRSS3; PRSS4; Serine protease 3; Serine protease 4; T9; Tb; Try3; TRY4; trypsin 3; trypsin III; Trypsin IV; trypsin-3; trypsinogen; trypsinogen 15; trypsinogen 4; trypsinogen 5; trypsinogen IV
Gene Symbols PRSS3
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Trypsin 3 Phe16-Asn246.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22073
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.