Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PRSS3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5103175
Description
Antibody detects endogenous levels of total Trypsin-3.
PRSS3 encodes a trypsinogen, which is a member of the trypsin family of serine proteases. This enzyme is expressed in the brain and pancreas and is resistant to common trypsin inhibitors. It is active on peptide linkages involving the carboxyl group of lysine or arginine. PRSS3 is localized to the locus of T cell receptor beta variable orphans on chromosome 9. Four transcript variants encoding different isoforms have been described for this gene.
Specifications
PRSS3 | |
Polyclonal | |
Unconjugated | |
PRSS3 | |
Brain trypsinogen; mesotrypsin; Mesotrypsinogen; MTG; pancreatic trypsinogen III; protease, serine 3; protease, serine, 3; protease, serine, 4 (trypsin 4, brain); PRSS3; PRSS4; Serine protease 3; Serine protease 4; T9; Tb; Try3; TRY4; trypsin 3; trypsin III; Trypsin IV; trypsin-3; trypsinogen; trypsinogen 15; trypsinogen 4; trypsinogen 5; trypsinogen IV | |
Rabbit | |
Affinity chromatography | |
RUO | |
22073, 5646 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P35030 | |
PRSS3 | |
A synthesized peptide derived from human PRSS3(Accession P35030), corresponding to amino acid residues P9-A59. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction