Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRSS35 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558542
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558542 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558542 Supplier Invitrogen™ Supplier No. PA558542
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MEWDFMWHLR KVPRIVSERT FHLTSPAFEA DAKMMVNTVC GIECQKELPT PSLSELEDYL SYETVFENGT RTLTRVKVQD LVLEPTQNIT TKGVSVRRK Highest antigen sequence identity to the following orthologs: Mouse - 67%, Rat - 70%.

The PRSS35 (Protease, Serine 35) gene encodes a protein that belongs to the trypsin-like serine protease family, which is involved in various physiological processes, including digestion, immune response, and blood coagulation. PRSS35 is located on chromosome 6 in humans. The protein is believed to play a role in extracellular matrix remodeling and may be involved in processes such as tissue repair and inflammation. Expression of PRSS35 has been detected in multiple tissues, including the reproductive system, where it may influence fertility and reproductive tract function. However, the specific substrates and detailed biological functions of PRSS35 are still under investigation. Alterations in PRSS35 expression have been associated with certain pathological conditions, such as cancer and inflammatory diseases, suggesting its potential as a biomarker and therapeutic target.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRSS35
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PRSS35
Gene Accession No. Q8N3Z0
Gene Alias 6030424L22Rik; C6orf158; dJ223E3.1; Inactive serine protease 35; P3D9; protease, serine 35; protease, serine, 35; PRSS35; UNQ522/PRO1057
Gene Symbols PRSS35
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PRSS35. Recombinant protein control fragment (Product #RP-94886).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 167681
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.